missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoimidazole Sulfate 98.0+%, TCI America™

Catalog No. p-7117569
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
A16651G 1 g
A16655G 5 g
2 options

Catalog No. A16651G

Supplier: TCI America A16651G

Only null left

Chemical Identifiers

CAS 1450-93-7
Molecular Formula C6H12N6O4S
Molecular Weight (g/mol) 264.26
MDL Number MFCD00013162
InChI Key KUWRLKJYNASPQZ-UHFFFAOYSA-N
Synonym 2-aminoimidazole hemisulfate, 1h-imidazol-2-amine sulfate 2:1, bis 1h-imidazol-2-amine ; sulfuric acid, 2-aminoimidazole sulphate, bis 2-aminoimidazole sulfate, 1h-imidazol-2-amine sulfurate, 2-aminoimidazole sulfate 2:1, 1h-imidazol-2-amine, sulfate 2:1, pubchem8378, aminoimidazole hemisulfate
PubChem CID 2734684
IUPAC Name bis(1H-imidazol-2-amine); sulfuric acid
SMILES OS(O)(=O)=O.NC1=NC=CN1.NC1=NC=CN1

Specifications

Melting Point 270°C
Quantity 1 g
Formula Weight 264.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoimidazole Sulfate