missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoimidazole sulfate, 98%

Catalog No. p-7028607
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2599903 1 g
AAH2599909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2599903 Supplier Thermo Scientific Chemicals Supplier No. H2599903

CAS: 1450-93-7 | C6H12N6O4S | 264.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1450-93-7
Molecular Formula C6H12N6O4S
Molecular Weight (g/mol) 264.26
MDL Number MFCD00013162
InChI Key KUWRLKJYNASPQZ-UHFFFAOYSA-N
Synonym 2-aminoimidazole hemisulfate, 1h-imidazol-2-amine sulfate 2:1, bis 1h-imidazol-2-amine ; sulfuric acid, 2-aminoimidazole sulphate, bis 2-aminoimidazole sulfate, 1h-imidazol-2-amine sulfurate, 2-aminoimidazole sulfate 2:1, 1h-imidazol-2-amine, sulfate 2:1, pubchem8378, aminoimidazole hemisulfate
PubChem CID 2734684
IUPAC Name 1H-imidazol-2-amine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=NC=CN1.NC1=NC=CN1

Specifications

Melting Point ∼270°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 4726750
Formula Weight 132.13
Percent Purity 98%
Chemical Name or Material 2-Aminoimidazole sulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-918-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only