missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoimidazole hemisulfate, 98+%, Thermo Scientific™

$159.66 - $530.18

Chemical Identifiers

CAS 1450-93-7
Molecular Formula 0·5 H2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00013162
InChI Key KUWRLKJYNASPQZ-UHFFFAOYSA-N
Synonym 2-aminoimidazole hemisulfate, 1h-imidazol-2-amine sulfate 2:1, bis 1h-imidazol-2-amine ; sulfuric acid, 2-aminoimidazole sulphate, bis 2-aminoimidazole sulfate, 1h-imidazol-2-amine sulfurate, 2-aminoimidazole sulfate 2:1, 1h-imidazol-2-amine, sulfate 2:1, pubchem8378, aminoimidazole hemisulfate
PubChem CID 2734684
IUPAC Name 1H-imidazol-2-amine;sulfuric acid
SMILES C1=CN=C(N1)N.C1=CN=C(N1)N.OS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC368860025
View Documents Promotion Details
N/A 2.5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC368860100
View Documents Promotion Details
N/A 10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1450-93-7
132.13
KUWRLKJYNASPQZ-UHFFFAOYSA-N
2734684
C1=CN=C(N1)N.C1=CN=C(N1)N.OS(=O)(=O)O
0·5 H2O4S
MFCD00013162
2-aminoimidazole hemisulfate, 1h-imidazol-2-amine sulfate 2:1, bis 1h-imidazol-2-amine ; sulfuric acid, 2-aminoimidazole sulphate, bis 2-aminoimidazole sulfate, 1h-imidazol-2-amine sulfurate, 2-aminoimidazole sulfate 2:1, 1h-imidazol-2-amine, sulfate 2:1, pubchem8378, aminoimidazole hemisulfate
1H-imidazol-2-amine;sulfuric acid

Specifications

1450-93-7
100.0
98+%
MFCD00013162
KUWRLKJYNASPQZ-UHFFFAOYSA-N
1H-imidazol-2-amine;sulfuric acid
2734684
98+%
98.0
Authentic
0·5 H2O4S
2-aminoimidazole hemisulfate, 1h-imidazol-2-amine sulfate 2:1, bis 1h-imidazol-2-amine ; sulfuric acid, 2-aminoimidazole sulphate, bis 2-aminoimidazole sulfate, 1h-imidazol-2-amine sulfurate, 2-aminoimidazole sulfate 2:1, 1h-imidazol-2-amine, sulfate 2:1, pubchem8378, aminoimidazole hemisulfate
C1=CN=C(N1)N.C1=CN=C(N1)N.OS(=O)(=O)O
132.13
132.13
2-Aminoimidazole hemisulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-918-9

SDS
Documents

Documents

RUO – Research Use Only