missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2-Aminoheptane

Catalog No. A56205100G
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A56205100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A56205100G Supplier Sigma Aldrich Supplier No. A56205100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Aminoheptane is a secondary alkyl amine that can be used to synthesize 3-amino-2-heptanone hydrochloride.

Specifications

CAS 123-82-0
Density 0.766 g/mL (at 25°C (literature))
Boiling Point 142°C to 144°C (lit.)
Molecular Formula C7H17N
Refractive Index n20/D 1.418 (literature)
Linear Formula CH3(CH2)4 CH(NH2)CH3
MDL Number MFCD00008101
Quantity 100 g
Synonym 1-Methylhexylamine; 2-Heptylamine
Molecular Weight (g/mol) 115.22
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less