missing translation for 'onlineSavingsMsg'
Learn More

(2-Aminoethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Catalog No. p-3756787
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291630010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC291630010 Supplier Thermo Scientific Chemicals Supplier No. 291630010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2041-14-7
Molecular Formula C2H8NO3P
Molecular Weight (g/mol) 125.06
MDL Number MFCD00008182
InChI Key QQVDJLLNRSOCEL-UHFFFAOYSA-N
Synonym 2-aminoethyl phosphonic acid, ciliatine, phosphonoethylamine, aminoethylphosphonic acid, 2-aminoethane phosphonic acid, 2-aminoethanephosphonic acid, 2-aminoethylphosphonate, beta-aminoethylphosphonic acid, 2-aminoethyl phosphonate, phosphonic acid, 2-aminoethyl
PubChem CID 339
ChEBI CHEBI:15573
IUPAC Name 2-aminoethylphosphonic acid
SMILES NCCP(O)(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 296.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NH2CH2CH2P(O)(OH)2
Quantity 1 g
Beilstein 04, 1792
Solubility Information Solubility in water: soluble in water. Other solubilities: insoluble in methanol and acetone
Formula Weight 125.06
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (2-Aminoethyl)phosphonic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-043-

RUO – Research Use Only