missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl Methyl Sulfone Hydrochloride 98.0+%, TCI America™

Catalog No. A24645G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24645G Supplier TCI America Supplier No. A24645G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104458-24-4
Molecular Formula C3H10ClNO2S
Molecular Weight (g/mol) 159.628
MDL Number MFCD03840162
InChI Key AMYYUKGKCJKCBI-UHFFFAOYSA-N
Synonym 2-(Methylsulfonyl)ethanamine Hydrochloride
PubChem CID 17749886
IUPAC Name 2-methylsulfonylethanamine;hydrochloride
SMILES CS(=O)(=O)CCN.Cl

Specifications

Melting Point 170°C
Color Yellow
Quantity 5 g
Formula Weight 159.63
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethyl Methyl Sulfone Hydrochloride