missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl Hydrogen Sulfate 98.0+%, TCI America™

Catalog No. p-7135888
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S044525G 25 g
S0445500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S044525G Supplier TCI America Supplier No. S044525G
Only null left

Chemical Identifiers

CAS 926-39-6
Molecular Formula C2H7NO4S
Molecular Weight (g/mol) 141.14
MDL Number MFCD00008179
InChI Key WSYUEVRAMDSJKL-UHFFFAOYSA-N
Synonym ethanolamine o-sulfate, aminoethyl sulfate, 2-aminoethyl sulfate, mono 2-aminoethyl sulfate, 2-aminoethyl hydrogen sulphate, sulfuric acid mono 2-aminoethyl ester, was-34, 2-aminoethylhydrogensulfate, unii-9c8f910hlk, ethanol, 2-amino-, hydrogen sulfate ester
PubChem CID 70223
IUPAC Name (2-aminoethoxy)sulfonic acid
SMILES NCCOS(O)(=O)=O

Specifications

Melting Point 279°C
Color White
Quantity 25 g
Formula Weight 141.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethyl Hydrogen Sulfate
EINECSNumber (2)-1676
RTECSNumber KJ6371000
TSCA Yes