missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl hydrogen sulfate, 99%

Catalog No. AC103645000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103645000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC103645000 Supplier Thermo Scientific Chemicals Supplier No. 103645000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 926-39-6 | C2H7NO4S | 141.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 926-39-6
Molecular Formula C2H7NO4S
Molecular Weight (g/mol) 141.14
MDL Number MFCD00008179
InChI Key WSYUEVRAMDSJKL-UHFFFAOYSA-N
Synonym ethanolamine o-sulfate, aminoethyl sulfate, 2-aminoethyl sulfate, mono 2-aminoethyl sulfate, 2-aminoethyl hydrogen sulphate, sulfuric acid mono 2-aminoethyl ester, was-34, 2-aminoethylhydrogensulfate, unii-9c8f910hlk, ethanol, 2-amino-, hydrogen sulfate ester
PubChem CID 70223
IUPAC Name 2-aminoethyl hydrogen sulfate
SMILES NCCOS(O)(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 277°C to 280°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NCH2CH2OSO3H
Quantity 500 g
Beilstein 04, 276
Solubility Information Solubility in water: soluble. Other solubilities: insolube in ethanol
Formula Weight 141.14
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Aminoethyl hydrogen sulfate, 99%
Show More Show Less
EINECSNumber 213-135-7
RTECSNumber KJ6371000
TSCA TSCA

RUO – Research Use Only