missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminoethyl Diphenylborinate 98.0+%, TCI America™
Supplier: TCI America D028110G
Specifications
| 2-Aminoethyl Diphenylborinate | |
| 190°C | |
| C14H16BNO | |
| 10 g | |
| BLZVCIGGICSWIG-UHFFFAOYSA-N | |
| 2-aminoethyl diphenylborinate | |
| 1598 | |
| ≥98.0% (HPLC,T) |
| 524-95-8 | |
| White | |
| MFCD00014823 | |
| 2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl | |
| NCCOB(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 225.10 | |
| 225.10 | |
| Crystalline Powder |
Chemical Identifiers
| 524-95-8 | |
| 225.10 | |
| BLZVCIGGICSWIG-UHFFFAOYSA-N | |
| 1598 | |
| NCCOB(C1=CC=CC=C1)C1=CC=CC=C1 |
| C14H16BNO | |
| MFCD00014823 | |
| 2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl | |
| 2-aminoethyl diphenylborinate |
Safety and Handling
RTECSNumber : ED6150000
TSCA : No