missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl Diphenylborinate 98.0+%, TCI America™

Catalog No. D028125G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D028125G 25 g
D028110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D028125G Supplier TCI America Supplier No. D028125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 524-95-8
Molecular Formula C14H16BNO
Molecular Weight (g/mol) 225.10
MDL Number MFCD00014823
InChI Key BLZVCIGGICSWIG-UHFFFAOYSA-N
Synonym 2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenyl
PubChem CID 1598
IUPAC Name 2-aminoethyl diphenylborinate
SMILES NCCOB(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 190°C
Color White
Quantity 25 g
Formula Weight 225.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethyl Diphenylborinate
RTECSNumber ED6150000
TSCA No