missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanol 99.0+%, TCI America™

Catalog No. A029725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A029725G 25 g
A0297500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A029725G Supplier TCI America Supplier No. A029725G

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
IUPAC Name 2-aminoethan-1-ol
SMILES NCCO

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 170°C
Quantity 25 g
UN Number 2491
Formula Weight 61.08
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Aminoethanol
EINECSNumber (2)-0301
RTECSNumber KJ5775000
TSCA Yes