missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanol Hydrochloride 98.0+%, TCI America™

Catalog No. p-7116814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
A029825G 25 g
A0298100G 100 g
A0298500G 500 g
3 options

Catalog No. A029825G

Supplier: TCI America A029825G

Only null left

Chemical Identifiers

CAS 2002-24-6
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.542
MDL Number MFCD00012892
InChI Key PMUNIMVZCACZBB-UHFFFAOYSA-N
Synonym 2-aminoethanol hydrochloride, ethanolamine hydrochloride, ethanolamine hcl, mea hydrochloride, monoethanolamine hydrochloride, ethanolamine chloride, colamine hydrochloride, ethanol, 2-amino-, hydrochloride, ethanolamine-hcl, 2-hydroxyethylammonium chloride
PubChem CID 74819
IUPAC Name 2-aminoethanol;hydrochloride
SMILES C(CO)N.Cl

Specifications

Melting Point 84°C
Color White
Quantity 25 g
Formula Weight 97.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethanol Hydrochloride
RTECSNumber KJ6370000
TSCA Yes