missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanesulfonic Acid 98.0+%, TCI America™

Catalog No. p-7116811
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A029525G 25 g
A0295500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A029525G Supplier TCI America Supplier No. A029525G
Only null left

Chemical Identifiers

CAS 107-35-7
Molecular Formula C2H7NO3S
Molecular Weight (g/mol) 125.14
MDL Number MFCD00008197
InChI Key XOAAWQZATWQOTB-UHFFFAOYSA-N
Synonym taurine, tauphon, ethanesulfonic acid, 2-amino, l-taurine, 2-aminoethylsulfonic acid, o-due, 2-sulfoethylamine, taufon, aminoethanesulfonic acid, aminoethylsulfonic acid
PubChem CID 1123
ChEBI CHEBI:15891
IUPAC Name 2-aminoethane-1-sulfonic acid
SMILES NCCS(O)(=O)=O

Specifications

Melting Point 300°C
Color White
Quantity 25 g
Formula Weight 125.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethanesulfonic Acid
EINECSNumber (2)-1607&(9)-0801
RTECSNumber WX0175000
TSCA Yes