missing translation for 'onlineSavingsMsg'
Learn More

2-Aminodiphenylamine 98.0+%, TCI America™

Catalog No. p-7117749
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
A21295G 5 g
A212925G 25 g
2 options

Catalog No. A21295G

Supplier: TCI America A21295G

Only null left

Chemical Identifiers

CAS 534-85-0
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00007685
InChI Key NFCPRRWCTNLGSN-UHFFFAOYSA-N
Synonym 2-aminodiphenylamine, n-phenyl-o-phenylenediamine, n1-phenylbenzene-1,2-diamine, o-semidine, 1,2-benzenediamine, n-phenyl, o-aminodiphenylamine, n-phenyl-1,2-phenylenediamine, n-phenylbenzene-1,2-diamine, o-phenylenediamine, n-phenyl, unii-g2x60k1y26
PubChem CID 68297
IUPAC Name N1-phenylbenzene-1,2-diamine
SMILES NC1=CC=CC=C1NC1=CC=CC=C1

Specifications

Melting Point 80°C
Boiling Point 108°C
Quantity 5 g
Formula Weight 184.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminodiphenylamine