missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobiphenyl 99.0+%, TCI America™

Catalog No. B046725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B046725G 25 g
B0467100G 100 g
B0467500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B046725G Supplier TCI America Supplier No. B046725G

Chemical Identifiers

CAS 90-41-5
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00007701
InChI Key TWBPWBPGNQWFSJ-UHFFFAOYSA-N
Synonym 2-aminobiphenyl, 1,1'-biphenyl-2-amine, 2-biphenylamine, 2-aminodiphenyl, o-aminobiphenyl, o-biphenylamine, biphenyl-2-ylamine, o-aminodiphenyl, o-phenylaniline, biphenyl-2-amine
PubChem CID 7015
IUPAC Name [1,1'-biphenyl]-2-amine
SMILES NC1=CC=CC=C1C1=CC=CC=C1

Specifications

Melting Point 51°C
Color White-Yellow
Boiling Point 299°C
Quantity 25 g
Formula Weight 169.23
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobiphenyl [for Sugar determination]
EINECSNumber (9)-0081
RTECSNumber DU8850000
TSCA Yes