missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobiphenyl, 98%

Catalog No. AC103220050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103220050 5 g Glass bottle
AC103220250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103220050 Supplier Thermo Scientific Chemicals Supplier No. 103220050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 90-41-5 | C12H11N | 169.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-41-5
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00007701
InChI Key TWBPWBPGNQWFSJ-UHFFFAOYSA-N
Synonym 2-aminobiphenyl, 1,1'-biphenyl-2-amine, 2-biphenylamine, 2-aminodiphenyl, o-aminobiphenyl, o-biphenylamine, biphenyl-2-ylamine, o-aminodiphenyl, o-phenylaniline, biphenyl-2-amine
PubChem CID 7015
IUPAC Name 2-phenylaniline
SMILES NC1=CC=CC=C1C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48°C to 52°C
Color Purple to Brown
Boiling Point 299°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H4NH2
Quantity 5 g
Beilstein 12, 1317
Formula Weight 169.23
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Aminobiphenyl, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-990-9
RTECSNumber DU8850000
TSCA TSCA

RUO – Research Use Only