missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzonitrile 98.0+%, TCI America™

Catalog No. A086125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A086125G 25 g
A0861100G 100 g
A0861500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A086125G Supplier TCI America Supplier No. A086125G

Chemical Identifiers

CAS 1885-29-6
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00007631
InChI Key HLCPWBZNUKCSBN-UHFFFAOYSA-N
Synonym anthranilonitrile, 2-cyanoaniline, o-aminobenzonitrile, benzonitrile, 2-amino, o-cyanoaniline, 2-amino-benzonitrile, aminobenzonitrile, aminobenzonitrile 2-, 2-aminobenzenecarbonitrile, unii-5rxh2b211r
PubChem CID 72913
IUPAC Name 2-aminobenzonitrile
SMILES NC1=CC=CC=C1C#N

Specifications

Melting Point 50°C
Boiling Point 263°C
Quantity 25 g
UN Number 3439
Formula Weight 118.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzonitrile
EINECSNumber (3)-1812
RTECSNumber CB4575000
TSCA Yes