missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzonitrile, 98%

Catalog No. p-7082043
Change view
Click to view available options
Quantity:
50 g
250 g
Catalog No. Quantity
AAA1228618 50 g
AAA1228630 250 g
2 options

Catalog No. AAA1228618

Supplier: Thermo Scientific Chemicals A1228618

Only null left

CAS: 1885-29-6 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1885-29-6
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00007631
InChI Key HLCPWBZNUKCSBN-UHFFFAOYSA-N
Synonym anthranilonitrile, 2-cyanoaniline, o-aminobenzonitrile, benzonitrile, 2-amino, o-cyanoaniline, 2-amino-benzonitrile, aminobenzonitrile, aminobenzonitrile 2-, 2-aminobenzenecarbonitrile, unii-5rxh2b211r
PubChem CID 72913
IUPAC Name 2-aminobenzonitrile
SMILES NC1=CC=CC=C1C#N

Specifications

Melting Point 45°C to 49°C
Boiling Point 286°C
Flash Point 145°C (293°F)
Odor Almond-like
Quantity 50 g
UN Number UN3439
Beilstein 907187
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 2-Aminobenzonitrile
Hazard Category H302+H312+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H311
Toxic in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 217-549-9
RTECSNumber CB4575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only