missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzimidazole 98.0+%, TCI America™

Catalog No. A08505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08505G 5 g
A085025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08505G Supplier TCI America Supplier No. A08505G

Chemical Identifiers

CAS 934-32-7
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005596
InChI Key JWYUFVNJZUSCSM-UHFFFAOYSA-N
Synonym 2-aminobenzimidazole, 1h-benzo d imidazol-2-amine, 2-iminobenzimidazoline, benzimidazole, 2-amino, benzimidazol-2-ylamine, 2-amino benzimidazole, caswell no. 033aa, 1h-1,3-benzodiazol-2-amine, usaf ek-4037, 2-amino-1h-benzimidazole
PubChem CID 13624
ChEBI CHEBI:27822
IUPAC Name 1H-1,3-benzodiazol-2-amine
SMILES NC1=NC2=CC=CC=C2N1

Specifications

Melting Point 231°C
Quantity 5 g
Formula Weight 133.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminobenzimidazole
EINECSNumber (5)-0462
RTECSNumber DD5775000
TSCA Yes