missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzimidazole, 97+%

Catalog No. p-7043686
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0206606 5 g
AAL0206614 25 g
AAL0206622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0206606 Supplier Thermo Scientific Chemicals Supplier No. L0206606

CAS: 934-32-7 | C7H7N3 | 133.15 g/mol

2-Aminobenzimidazole is used as receptors in chemosensor analysis. It was used in the hydrolysis of a choline carbonate. It was also used in the synthesis of imidazo[1,2-a]benzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzimidazole is used as receptors in chemosensor analysis. It was used in the hydrolysis of a choline carbonate. It was also used in the synthesis of imidazo[1,2-a]benzimidazoles.

Solubility
Soluble in water (Slightly).

Notes
Protect from direct light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 934-32-7
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005596
InChI Key JWYUFVNJZUSCSM-UHFFFAOYSA-N
Synonym 2-aminobenzimidazole, 1h-benzo d imidazol-2-amine, 2-iminobenzimidazoline, benzimidazole, 2-amino, benzimidazol-2-ylamine, 2-amino benzimidazole, caswell no. 033aa, 1h-1,3-benzodiazol-2-amine, usaf ek-4037, 2-amino-1h-benzimidazole
PubChem CID 13624
ChEBI CHEBI:27822
IUPAC Name 1H-benzimidazol-2-amine
SMILES NC1=NC2=CC=CC=C2N1

Specifications

Melting Point 228°C to 232°C
Quantity 5 g
Beilstein 116525
Solubility Information Soluble in water (Slightly).
Formula Weight 133.15
Percent Purity ≥97%
Chemical Name or Material 2-Aminobenzimidazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H361-H315-H319-H335
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 213-280-6
RTECSNumber DD5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only