missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzhydrazide, 98+%

Catalog No. AAA1080509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1080509 10 g
AAA1080518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1080509 Supplier Thermo Scientific Chemicals Supplier No. A1080509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1904-58-1 | C7H9N3O | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1904-58-1
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.17
MDL Number MFCD00017056
InChI Key GRWMSCBKWMQPON-UHFFFAOYSA-N
Synonym 2-aminobenzhydrazide, o-aminobenzhydrazide, anthranilic acid hydrazide, benzoic acid, 2-amino-, hydrazide, anthranilic hydrazide, anthraniloyl hydrazide, anthraniloyl hydrazine, 2-aminobenzoylhydrazine, 2-aminobenzoic acid hydrazide, o-aminobenzoic hydrazide
PubChem CID 94728
IUPAC Name 2-aminobenzohydrazide
SMILES NNC(=O)C1=CC=CC=C1N

Specifications

Melting Point 122°C to 124°C
Quantity 10 g
Beilstein 742811
Formula Weight 151.17
Percent Purity ≥98%
Chemical Name or Material 2-Aminobenzhydrazide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 217-604-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only