missing translation for 'onlineSavingsMsg'
Learn More

2'-Aminoacetophenone Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022602
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022602 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022602 Supplier Merck Emd Millipore Supplier No. 49868250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2'-Aminoacetophenone is a volatile aromatic compound, which is typically used as a key flavor ingredient in food and beverages.

Specifications

CAS 551-93-9
Density 1.112 g/mL (at 25°C (literature))
Boiling Point 85°C to 90°C/0.5 mmHg (literature)
Flash Point 113°C (Closed Cup)
Molecular Formula C8H9NO
Refractive Index n20/D 1.613-1.619; n20/D 1.614 (literature)
Linear Formula H2NC6H4COCH3
Quantity 250 mg
MDL Number MFCD00007717
UN Number NONH for all modes of transport
Synonym 2-Acetylaniline
Formula Weight 135.16
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less