missing translation for 'onlineSavingsMsg'
Learn More

2'-Aminoacetophenone, 98%

Catalog No. AAA1089506
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1089506 5 g
AAA1089518 50 g
AAA1089530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1089506 Supplier Thermo Scientific Chemicals Supplier No. A1089506

CAS: 551-93-9 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 551-93-9
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007717
InChI Key GTDQGKWDWVUKTI-UHFFFAOYSA-N
Synonym 2'-aminoacetophenone, 1-2-aminophenyl ethanone, o-aminoacetophenone, 2-acetylaniline, o-acetylaniline, o-aminoacetylbenzene, 1-acetyl-2-aminobenzene, 1-2-aminophenyl ethan-1-one, acetophenone, 2'-amino, ethanone, 1-2-aminophenyl
PubChem CID 11086
IUPAC Name 1-(2-aminophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1N

Specifications

Density 1.118
Boiling Point 253°C to 255°C
Flash Point 112°C (233°F)
Refractive Index 1.616
Quantity 5 g
Beilstein 386122
Merck Index 14,413
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material 2'-Aminoacetophenone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-002-8
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only