missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-p-cresol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 95-84-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00007699
InChI Key ZMXYNJXDULEQCK-UHFFFAOYSA-N
Synonym 2-amino-p-cresol, 2-hydroxy-5-methylaniline, phenol, 2-amino-4-methyl, 3-amino-4-hydroxytoluene, 6-hydroxy-m-toluidine, 4-methyl-2-aminophenol, 5-methyl-2-hydroxyaniline, 2-amino-4-cresol, p-cresol, 2-amino, 2-amino-4-methyl-phenol
PubChem CID 7264
IUPAC Name 2-amino-4-methylphenol
SMILES CC1=CC=C(O)C(N)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC163210500
SDS View Documents
Thermo Scientific Chemicals
163210500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC163212500
SDS View Documents
Thermo Scientific Chemicals
163212500
250g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

95-84-1
123.16
ZMXYNJXDULEQCK-UHFFFAOYSA-N
7264
CC1=CC=C(O)C(N)=C1
C7H9NO
MFCD00007699
2-amino-p-cresol, 2-hydroxy-5-methylaniline, phenol, 2-amino-4-methyl, 3-amino-4-hydroxytoluene, 6-hydroxy-m-toluidine, 4-methyl-2-aminophenol, 5-methyl-2-hydroxyaniline, 2-amino-4-cresol, p-cresol, 2-amino, 2-amino-4-methyl-phenol
2-amino-4-methylphenol

Specifications

95-84-1
100.0
96% min. (GC)
C7H9NO
MFCD00007699
13,601
Solubility in water: slightly soluble
CC1=CC=C(O)C(N)=C1
123.16
123.16
2-Amino-p-cresol, 97%
96.0
Authentic
Glass bottle
H2NC6H3(CH3)OH
50g
2-amino-p-cresol, 2-hydroxy-5-methylaniline, phenol, 2-amino-4-methyl, 3-amino-4-hydroxytoluene, 6-hydroxy-m-toluidine, 4-methyl-2-aminophenol, 5-methyl-2-hydroxyaniline, 2-amino-4-cresol, p-cresol, 2-amino, 2-amino-4-methyl-phenol
ZMXYNJXDULEQCK-UHFFFAOYSA-N
2-amino-4-methylphenol
7264
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-457-3

RTECSNumber : SJ6078000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only