missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride, 97%

Catalog No. p-7035675
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5339803 1 g
AAH5339806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5339803 Supplier Thermo Scientific Chemicals Supplier No. H5339803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1171331-39-7 | C4H8ClF3N2O | 192.566 g/mol

Reactant in the synthesis of isoxazoline indolizine amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of isoxazoline indolizine amides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1171331-39-7
Molecular Formula C4H8ClF3N2O
Molecular Weight (g/mol) 192.566
MDL Number MFCD09944516
InChI Key DBNFKWRZLGVLSH-UHFFFAOYSA-N
Synonym 2-amino-n-2,2,2-trifluoroethyl acetamide hydrochloride, 2-amino-n-2,2,2-trifluoroethyl acetamide;hydrochloride, c4h7f3n2o.hcl, 2-amino-n-2,2,2-trifluoroethyl acetamide hcl
PubChem CID 42913698
IUPAC Name 2-amino-N-(2,2,2-trifluoroethyl)acetamide;hydrochloride
SMILES C(C(=O)NCC(F)(F)F)N.Cl

Specifications

Linear Formula H2NCH2(CO)NHCH2CF3·HCl
Quantity 1 g
Formula Weight 192.57
Percent Purity 97%
Chemical Name or Material 2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only