missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene 98.0+%, TCI America™

Catalog No. A28585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28585G 5 g
A28581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28585G Supplier TCI America Supplier No. A28585G

Chemical Identifiers

CAS 1246562-40-2
Molecular Formula C33H26BrN
Molecular Weight (g/mol) 516.48
MDL Number MFCD20040455
InChI Key CBLKFNHDNANUNU-UHFFFAOYSA-N
PubChem CID 58131689
IUPAC Name N-{[1,1'-biphenyl]-4-yl}-N-(4-bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine
SMILES CC1(C)C2=CC=CC=C2C2=C1C=C(C=C2)N(C1=CC=C(Br)C=C1)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 5 g
Formula Weight 516.48
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-N-[(1,1′-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene