missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-(trifluoromethyl)pyridine 98.0+%, TCI America™
$42.36 - $150.16
Chemical Identifiers
| CAS | 34486-24-3 |
|---|---|
| Molecular Formula | C6H5F3N2 |
| Molecular Weight (g/mol) | 162.115 |
| MDL Number | MFCD03093868 |
| InChI Key | NFYYDQMFURPHCC-UHFFFAOYSA-N |
| Synonym | 2-amino-6-trifluoromethyl pyridine, 6-trifluoromethyl pyridin-2-amine, 2-amino-6-trifluoromethylpyridine, 6-trifluoromethyl-2-pyridinamine, 6-trifluoromethyl-2-pyridinylamine, 6-trifluoromethyl-2-pyridylamine, 6-trifluoromethyl-pyridin-2-ylamine, 2-pyridinamine, 6-trifluoromethyl, acmc-209i7w |
| PubChem CID | 821024 |
| IUPAC Name | 6-(trifluoromethyl)pyridin-2-amine |
| SMILES | C1=CC(=NC(=C1)N)C(F)(F)F |
Chemical Identifiers
| 34486-24-3 | |
| 162.115 | |
| NFYYDQMFURPHCC-UHFFFAOYSA-N | |
| 821024 | |
| C1=CC(=NC(=C1)N)C(F)(F)F |
| C6H5F3N2 | |
| MFCD03093868 | |
| 2-amino-6-trifluoromethyl pyridine, 6-trifluoromethyl pyridin-2-amine, 2-amino-6-trifluoromethylpyridine, 6-trifluoromethyl-2-pyridinamine, 6-trifluoromethyl-2-pyridinylamine, 6-trifluoromethyl-2-pyridylamine, 6-trifluoromethyl-pyridin-2-ylamine, 2-pyridinamine, 6-trifluoromethyl, acmc-209i7w | |
| 6-(trifluoromethyl)pyridin-2-amine |
Specifications
| 34486-24-3 | |
| C6H5F3N2 | |
| 1 g | |
| 2-amino-6-trifluoromethyl pyridine, 6-trifluoromethyl pyridin-2-amine, 2-amino-6-trifluoromethylpyridine, 6-trifluoromethyl-2-pyridinamine, 6-trifluoromethyl-2-pyridinylamine, 6-trifluoromethyl-2-pyridylamine, 6-trifluoromethyl-pyridin-2-ylamine, 2-pyridinamine, 6-trifluoromethyl, acmc-209i7w | |
| C1=CC(=NC(=C1)N)C(F)(F)F | |
| 162.115 | |
| 162.12 | |
| Crystalline Powder |
| 87°C | |
| MFCD03093868 | |
| 2811 | |
| NFYYDQMFURPHCC-UHFFFAOYSA-N | |
| 6-(trifluoromethyl)pyridin-2-amine | |
| 821024 | |
| ≥98.0% (GC,T) | |
| 2-Amino-6-(trifluoromethyl)pyridine |
Safety and Handling
TSCA : No