missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-(trifluoromethyl)pyridine, 98+%

Catalog No. p-7047558
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
500 mg
Catalog No. Quantity
AAL19443MC 100 mg
AAL19443ME 500 mg
2 options

Catalog No. AAL19443MC

Supplier: Thermo Scientific Chemicals L19443MC

Only null left

CAS: 34486-24-3 | C6H5F3N2 | 162.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34486-24-3
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.115
MDL Number MFCD03093868
InChI Key NFYYDQMFURPHCC-UHFFFAOYSA-N
Synonym 2-amino-6-trifluoromethyl pyridine, 6-trifluoromethyl pyridin-2-amine, 2-amino-6-trifluoromethylpyridine, 6-trifluoromethyl-2-pyridinamine, 6-trifluoromethyl-2-pyridinylamine, 6-trifluoromethyl-2-pyridylamine, 6-trifluoromethyl-pyridin-2-ylamine, 2-pyridinamine, 6-trifluoromethyl, acmc-209i7w
PubChem CID 821024
IUPAC Name 6-(trifluoromethyl)pyridin-2-amine
SMILES C1=CC(=NC(=C1)N)C(F)(F)F

Specifications

Melting Point 86°C to 89°C
Quantity 100 mg
UN Number UN2811
Beilstein 473259
Formula Weight 162.11
Percent Purity ≥98%
Chemical Name or Material 2-Amino-6-(trifluoromethyl)pyridine
Hazard Category H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only