missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-(trifluoromethyl)benzothiazole 98.0+%, TCI America™

Catalog No. p-7117923
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24611G 1 g
A24615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24611G Supplier TCI America Supplier No. A24611G

Chemical Identifiers

CAS 777-12-8
Molecular Formula C8H5F3N2S
Molecular Weight (g/mol) 218.197
MDL Number MFCD00269597
InChI Key WEDYEBJLWMPPOK-UHFFFAOYSA-N
Synonym 2-amino-6-trifluoromethyl benzothiazole, 6-trifluoromethyl benzo d thiazol-2-amine, 6-trifluoromethyl-1,3-benzothiazol-2-amine, 2-amino-6-trifluoromethyl-1,3-benzothiazole, 2-amino-6-trifluoromethylbenzothiazole, 6-trifluoromethyl-benzothiazol-2-ylamine, 2-benzothiazolamine, 6-trifluoromethyl, 2-amino-6-trifluoromethyl-1,3-benzothiazol, 6-trifluoromethyl-1,3-benzothiazol-2-ylamine, 6-trifluoromethyl benzothiazole-2-ylamine
PubChem CID 2735955
IUPAC Name 6-(trifluoromethyl)-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1C(F)(F)F)SC(=N2)N

Specifications

Melting Point 116°C
Color Yellow
Quantity 1 g
Formula Weight 218.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-(trifluoromethyl)benzothiazole