missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylpyridine 98.0+%, TCI America™

Catalog No. p-7116879
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A040325G 25 g
A0403100G 100 g
A0403500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A040325G Supplier TCI America Supplier No. A040325G
Only null left

Chemical Identifiers

CAS 1824-81-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006331
InChI Key QUXLCYFNVNNRBE-UHFFFAOYSA-N
Synonym 2-amino-6-methylpyridine, 2-amino-6-picoline, 6-amino-2-picoline, 2-pyridinamine, 6-methyl, 6-methyl-2-pyridinamine, 6-methyl-2-pyridylamine, 6-methyl-2-aminopyridine, 2-picoline, 6-amino, 6-amino-2-methylpyridine, pyridine, 2-amino-6-methyl
PubChem CID 15765
IUPAC Name 6-methylpyridin-2-amine
SMILES CC1=CC=CC(N)=N1

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 209°C
Quantity 25 g
UN Number 2811
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Amino-6-methylpyridine
EINECSNumber (9)-0117
RTECSNumber US1885000
TSCA Yes