missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylpyridine, 99%

Catalog No. p-7080769
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1044422 100 g
AAA1044436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1044422 Supplier Thermo Scientific Chemicals Supplier No. A1044422
Only null left

CAS: 1824-81-3 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1824-81-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006331
InChI Key QUXLCYFNVNNRBE-UHFFFAOYSA-N
Synonym 2-amino-6-methylpyridine, 2-amino-6-picoline, 6-amino-2-picoline, 2-pyridinamine, 6-methyl, 6-methyl-2-pyridinamine, 6-methyl-2-pyridylamine, 6-methyl-2-aminopyridine, 2-picoline, 6-amino, 6-amino-2-methylpyridine, pyridine, 2-amino-6-methyl
PubChem CID 15765
IUPAC Name 6-methylpyridin-2-amine
SMILES CC1=CC=CC(N)=N1

Specifications

Melting Point 41°C to 45°C
Boiling Point 208°C to 209°C
Flash Point 103°C (217°F)
Odor Unpleasant
Quantity 100 g
UN Number UN2811
Beilstein 107048
Sensitivity Hygroscopic
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2-Amino-6-methylpyridine
Show More Show Less
Hazard Category H301-H310-H315-H319-H335
Hazard Statement GHS H Statement
H301-H310-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 217-360-1
RTECSNumber US1885000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only