missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylbenzothiazole 98.0+%, TCI America™

Catalog No. p-7117019
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A071425G 25 g
A0714100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A071425G Supplier TCI America Supplier No. A071425G
Only null left

Chemical Identifiers

CAS 2536-91-6
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00005789
InChI Key DZWTXWPRWRLHIL-UHFFFAOYSA-N
Synonym 2-amino-6-methylbenzothiazole, 6-methylbenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methyl, 6-methyl-2-aminobenzothiazole, 2-methylbenzothiazol-2-amine, 6-methyl-2-benzothiazolylamine, benzothiazole, 2-amino-6-methyl, 6-methyl-benzothiazol-2-ylamine, 2-amino-6-methyl-benzothiazole, 6-methyl-2-benzothiazolamine
PubChem CID 17335
IUPAC Name 6-methyl-1,3-benzothiazol-2-amine
SMILES CC1=CC=C2N=C(N)SC2=C1

Specifications

Melting Point 138°C
Color Yellow
Quantity 25 g
Formula Weight 164.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methylbenzothiazole
EINECSNumber (5)-0235
RTECSNumber DL2625000
TSCA Yes