missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylbenzothiazole, 99%

Catalog No. AAB2342214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2342214 25 g
AAB2342222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2342214 Supplier Thermo Scientific Chemicals Supplier No. B2342214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2536-91-6 | C8H8N2S | 164.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2536-91-6
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00005789
InChI Key DZWTXWPRWRLHIL-UHFFFAOYSA-N
Synonym 2-amino-6-methylbenzothiazole, 6-methylbenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methyl, 6-methyl-2-aminobenzothiazole, 2-methylbenzothiazol-2-amine, 6-methyl-2-benzothiazolylamine, benzothiazole, 2-amino-6-methyl, 6-methyl-benzothiazol-2-ylamine, 2-amino-6-methyl-benzothiazole, 6-methyl-2-benzothiazolamine
PubChem CID 17335
IUPAC Name 6-methyl-1,3-benzothiazol-2-amine
SMILES CC1=CC=C2N=C(N)SC2=C1

Specifications

Melting Point 139°C to 142°C
Quantity 25 g
Formula Weight 164.22
Percent Purity 99%
Chemical Name or Material 2-Amino-6-methylbenzothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 219-801-3
RTECSNumber DL2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only