missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxypyridine 98.0+%, TCI America™

Catalog No. p-7117796
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22281G 1 g
A22285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22281G Supplier TCI America Supplier No. A22281G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17920-35-3 | C6H8N2O | 124.143 g/mol

Chemical Identifiers

CAS 17920-35-3
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD04972542
InChI Key DEUALFRBMNMGDS-UHFFFAOYSA-N
Synonym 2-amino-6-methoxypyridine, 6-methoxy-pyridin-2-ylamine, 6-methoxy-2-pyridinamine, 6-methoxypyrid-2-ylamine, 6-methoxy-2-pyridylamine, 2-amino-6-methoxy pyridine, 2-pyridinamine, 6-methoxy, methoxypyridin-6-amine, acmc-209efc, 6-methoxy-2-aminopyridin
PubChem CID 580053
IUPAC Name 6-methoxypyridin-2-amine
SMILES COC1=CC=CC(=N1)N

Specifications

Boiling Point 115°C
Quantity 1 g
Formula Weight 124.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Amino-6-methoxypyridine
TSCA No
Recommended Storage Refrigerator