missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-methoxybenzothiazole 98.0+%, TCI America™
$37.29 - $277.75
Chemical Identifiers
| CAS | 1747-60-0 |
|---|---|
| Molecular Formula | C8H8N2OS |
| Molecular Weight (g/mol) | 180.225 |
| MDL Number | MFCD00005787 |
| InChI Key | KZHGPDSVHSDCMX-UHFFFAOYSA-N |
| Synonym | 2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393 |
| PubChem CID | 15630 |
| IUPAC Name | 6-methoxy-1,3-benzothiazol-2-amine |
| SMILES | COC1=CC2=C(C=C1)N=C(S2)N |
Chemical Identifiers
| 1747-60-0 | |
| 180.225 | |
| KZHGPDSVHSDCMX-UHFFFAOYSA-N | |
| 15630 | |
| COC1=CC2=C(C=C1)N=C(S2)N |
| C8H8N2OS | |
| MFCD00005787 | |
| 2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393 | |
| 6-methoxy-1,3-benzothiazol-2-amine |
Specifications
| 1747-60-0 | |
| C8H8N2OS | |
| 25 g | |
| 2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393 | |
| COC1=CC2=C(C=C1)N=C(S2)N | |
| 180.225 | |
| 180.23 | |
| Crystalline Powder |
| 168°C | |
| MFCD00005787 | |
| 2811 | |
| KZHGPDSVHSDCMX-UHFFFAOYSA-N | |
| 6-methoxy-1,3-benzothiazol-2-amine | |
| 15630 | |
| ≥98.0% (T) | |
| 2-Amino-6-methoxybenzothiazole |
Safety and Handling
EINECSNumber : (5)-0238
RTECSNumber : DL2100000
TSCA : Yes