missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxybenzothiazole 98.0+%, TCI America™

Catalog No. A071525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A071525G 25 g
A0715100G 100 g
A0715500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A071525G Supplier TCI America Supplier No. A071525G

Chemical Identifiers

CAS 1747-60-0
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.225
MDL Number MFCD00005787
InChI Key KZHGPDSVHSDCMX-UHFFFAOYSA-N
Synonym 2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393
PubChem CID 15630
IUPAC Name 6-methoxy-1,3-benzothiazol-2-amine
SMILES COC1=CC2=C(C=C1)N=C(S2)N

Specifications

Melting Point 168°C
Quantity 25 g
UN Number 2811
Formula Weight 180.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methoxybenzothiazole
EINECSNumber (5)-0238
RTECSNumber DL2100000
TSCA Yes