missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-methoxybenzothiazole, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 1747-60-0 |
|---|---|
| Molecular Formula | C8H8N2OS |
| Molecular Weight (g/mol) | 180.23 |
| MDL Number | MFCD00005787 |
| InChI Key | KZHGPDSVHSDCMX-UHFFFAOYSA-N |
| Synonym | 2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393 |
| PubChem CID | 15630 |
| IUPAC Name | 6-methoxy-1,3-benzothiazol-2-amine |
| SMILES | COC1=CC2=C(C=C1)N=C(S2)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC172020050
|
Thermo Scientific Chemicals
172020050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC172020500
|
Thermo Scientific Chemicals
172020500 |
50g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1747-60-0 | |
| 180.23 | |
| KZHGPDSVHSDCMX-UHFFFAOYSA-N | |
| 15630 | |
| COC1=CC2=C(C=C1)N=C(S2)N |
| C8H8N2OS | |
| MFCD00005787 | |
| 2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393 | |
| 6-methoxy-1,3-benzothiazol-2-amine |
Specifications
| 1747-60-0 | |
| 100.0 | |
| 98% | |
| C8H8N2OS | |
| 5g | |
| 2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393 | |
| COC1=CC2=C(C=C1)N=C(S2)N | |
| 180.23 | |
| 180.23 | |
| 2-Amino-6-methoxybenzothiazole, 98% |
| 97.5 | |
| Authentic | |
| Glass bottle | |
| MFCD00005787 | |
| 27, II, 334 | |
| KZHGPDSVHSDCMX-UHFFFAOYSA-N | |
| 6-methoxy-1,3-benzothiazol-2-amine | |
| 15630 | |
| 98% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Danger
EINECSNumber : 217-130-0
RTECSNumber : DL2100000
TSCA : TSCA
RUO â Research Use Only