missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxybenzothiazole, 98%

Catalog No. p-7026977
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2338009 10 g
AAB2338018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2338009 Supplier Thermo Scientific Chemicals Supplier No. B2338009
Only null left

CAS: 1747-60-0 | C8H8N2OS | 180.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1747-60-0
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.225
MDL Number MFCD00005787
InChI Key KZHGPDSVHSDCMX-UHFFFAOYSA-N
Synonym 2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393
PubChem CID 15630
IUPAC Name 6-methoxy-1,3-benzothiazol-2-amine
SMILES COC1=CC2=C(C=C1)N=C(S2)N

Specifications

Melting Point 161°C to 164°C
Odor Odorless
Quantity 10 g
Formula Weight 180.23
Percent Purity 98%
Chemical Name or Material 2-Amino-6-methoxybenzothiazole
Hazard Category H301-H341
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 217-130-0
RTECSNumber DL2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only