missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-iodopurine 98.0+%, TCI America™

Catalog No. p-7117710
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20685G 5 g
A206825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20685G Supplier TCI America Supplier No. A20685G

Chemical Identifiers

CAS 19690-23-4
Molecular Formula C5H4IN5
Molecular Weight (g/mol) 261.03
MDL Number MFCD03840496,MFCD08706375
InChI Key CQYPNVKLVHHOSJ-UHFFFAOYSA-N
Synonym 2-amino-6-iodopurine, 6-iodo-9h-purin-2-amine, 2-amino-6-iodo purine, 2-amino-6-iodo-7h-purine, pubchem9213, 6-iodo-2-aminopurine, 2-amino-6-iodo-purine, 6-iodopurine-2-ylamine, acmc-1caa5, amtns010
PubChem CID 6101575
IUPAC Name 6-iodo-7H-purin-2-amine
SMILES NC1=NC(I)=C2NC=NC2=N1

Specifications

Melting Point 240°C
Color Yellow
Quantity 5 g
Formula Weight 261.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-iodopurine