missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-fluorobenzothiazole, 99%

Catalog No. AAA1838303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1838303 1 g
AAA1838306 5 g
AAA1838314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1838303 Supplier Thermo Scientific Chemicals Supplier No. A1838303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 348-40-3 | C7H5FN2S | 168.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 348-40-3
Molecular Formula C7H5FN2S
Molecular Weight (g/mol) 168.189
MDL Number MFCD00013336
InChI Key CJLUXPZQUXVJNF-UHFFFAOYSA-N
Synonym 2-amino-6-fluorobenzothiazole, 6-fluorobenzo d thiazol-2-amine, 6-fluoro-benzothiazol-2-ylamine, 2-benzothiazolamine, 6-fluoro, 6-fluoro-2-benzothiazolamine, 2-amino-6-fluoro benzothiazole, 2-amino-6-fluoro-1,3-benzothiazole, 6-fluorobenzothiazole-2-ylamine, pubchem14467, maybridge1_006546
PubChem CID 319954
IUPAC Name 6-fluoro-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1F)SC(=N2)N

Specifications

Melting Point 183°C to 185°C
Quantity 1 g
Formula Weight 168.19
Percent Purity 99%
Chemical Name or Material 2-Amino-6-fluorobenzothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only