missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-fluorobenzoic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 434-76-4
Molecular Formula C7H5FNO2
Molecular Weight (g/mol) 154.12
MDL Number MFCD00067781
InChI Key RWSFZKWMVWPDGZ-UHFFFAOYSA-M
Synonym 6-fluoroanthranilic acid, anthralic acid, 6-fluoro, benzoic acid, 2-amino-6-fluoro, 6-amino-2-fluorobenzoic acid, 2-amino-6-fluoro-benzoic acid, 2,amino-6-fluorobenzoic acid, 4owo, pubchem1324, 6-fluoroanthanilic acid, acmc-209jun
PubChem CID 521142
IUPAC Name 2-amino-6-fluorobenzoate
SMILES NC1=CC=CC(F)=C1C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC306450050
SDS View Documents
Thermo Scientific Chemicals
306450050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

434-76-4
154.12
RWSFZKWMVWPDGZ-UHFFFAOYSA-M
521142
NC1=CC=CC(F)=C1C([O-])=O
C7H5FNO2
MFCD00067781
6-fluoroanthranilic acid, anthralic acid, 6-fluoro, benzoic acid, 2-amino-6-fluoro, 6-amino-2-fluorobenzoic acid, 2-amino-6-fluoro-benzoic acid, 2,amino-6-fluorobenzoic acid, 4owo, pubchem1324, 6-fluoroanthanilic acid, acmc-209jun
2-amino-6-fluorobenzoate

Specifications

434-76-4
100.0
97.5% min. (HPLC)
C7H5FNO2
MFCD00067781
6-fluoroanthranilic acid, anthralic acid, 6-fluoro, benzoic acid, 2-amino-6-fluoro, 6-amino-2-fluorobenzoic acid, 2-amino-6-fluoro-benzoic acid, 2,amino-6-fluorobenzoic acid, 4owo, pubchem1324, 6-fluoroanthanilic acid, acmc-209jun
NC1=CC=CC(F)=C1C([O-])=O
154.12
155.13
2-Amino-6-fluorobenzoic acid
97.5
Authentic
Glass bottle
H2NC6H3(F)COOH
5g
RWSFZKWMVWPDGZ-UHFFFAOYSA-M
2-amino-6-fluorobenzoate
521142
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only