missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropyrazine 98.0+%, TCI America™

Catalog No. p-7117757
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
A21451G 1 g
A21455G 5 g
2 options

Catalog No. A21451G

Supplier: TCI America A21451G

Only null left

Chemical Identifiers

CAS 33332-28-4
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD00055024
InChI Key JTPXVCKCLBROOJ-UHFFFAOYSA-N
Synonym 2-amino-6-chloropyrazine, 2-chloro-6-aminopyrazine, 2-pyrazinamine, 6-chloro, pyrazinamine, 6-chloro, 6-chloro-pyrazin-2-ylamine, 6-chloro-2-pyrazinamine, 6-chloropyrazin-2-ylamine, 2-amino-6-chloro-pyrazine, 6-chloropyrazine-2-ylamine, 6-amino-2-chloropyrazine
PubChem CID 118458
IUPAC Name 6-chloropyrazin-2-amine
SMILES C1=C(N=C(C=N1)Cl)N

Specifications

Melting Point 153°C
Quantity 1 g
Formula Weight 129.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-chloropyrazine