missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropurine 98.0+%, TCI America™

Catalog No. p-7117416
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A14071G 1 g
A14075G 5 g
2 options

Catalog No. A14071G

Supplier: TCI America A14071G

Only null left

Chemical Identifiers

CAS 10310-21-1
Molecular Formula C5H4ClN5
Molecular Weight (g/mol) 169.57
MDL Number MFCD00075252
InChI Key RYYIULNRIVUMTQ-UHFFFAOYSA-N
Synonym 2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine
PubChem CID 5360349
ChEBI CHEBI:72345
IUPAC Name 6-chloro-7H-purin-2-amine
SMILES NC1=NC(Cl)=C2NC=NC2=N1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 169.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-chloropurine
EINECSNumber (5)-6441
RTECSNumber UO7502000
TSCA No