missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropurine 98.0+%, TCI America™

Catalog No. A14075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14075G 5 g
A14071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14075G Supplier TCI America Supplier No. A14075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10310-21-1
Molecular Formula C5H4ClN5
Molecular Weight (g/mol) 169.57
MDL Number MFCD00075252
InChI Key RYYIULNRIVUMTQ-UHFFFAOYSA-N
Synonym 2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine
PubChem CID 5360349
ChEBI CHEBI:72345
IUPAC Name 6-chloro-7H-purin-2-amine
SMILES NC1=NC(Cl)=C2NC=NC2=N1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 169.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-chloropurine
EINECSNumber (5)-6441
RTECSNumber UO7502000
TSCA No