missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropurine, 99%

Catalog No. aaa1819506
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1819506 5 g
AAA1819503 1 g
AAA1819514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1819506 Supplier Thermo Scientific Chemicals Supplier No. A1819506
Only null left

CAS: 10310-21-1 | C5H4ClN5 | 169.57 g/mol

2-Amino-6-chloropurine finds its uses in the enzymatic synthesis of 2?-deoxyguanosine, 9-alkyl purines and (R)- and (S)-N-(2-phosphonomethoxypropyl) derivatives of purine and pyrimidine bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chloropurine finds its uses in the enzymatic synthesis of 2′-deoxyguanosine, 9-alkyl purines and (R)- and (S)-N-(2-phosphonomethoxypropyl) derivatives of purine and pyrimidine bases.

Solubility
Solubility in 1mol/L NaOH; very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 10310-21-1
Molecular Formula C5H4ClN5
Molecular Weight (g/mol) 169.57
MDL Number MFCD00075252
InChI Key RYYIULNRIVUMTQ-UHFFFAOYSA-N
Synonym 2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine
PubChem CID 5360349
ChEBI CHEBI:72345
IUPAC Name 6-chloro-7H-purin-2-amine
SMILES NC1=NC(Cl)=C2NC=NC2=N1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 9626
Formula Weight 169.57
Percent Purity 99%
Chemical Name or Material 2-Amino-6-chloropurine
Hazard Category H315-H319-H335-H360
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 233-686-7
RTECSNumber UO7502000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only