missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzonitrile, 98%

Catalog No. AAL0688303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0688303 1 g
AAL0688306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0688303 Supplier Thermo Scientific Chemicals Supplier No. L0688303

CAS: 6575-11-7 | C7H5ClN2 | 152.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6575-11-7
MDL Number MFCD00015546

Specifications

Melting Point 133°C to 137°C
Quantity 1 g
Molecular Formula C15H15BrClN2O4P
UN Number UN3439
Beilstein 2717090
Synonym 6-Chloroanthranilonitrile
InChI Key QEIFSLUFHRCVQL-UHFFFAOYSA-N
Molecular Weight (g/mol) 433.62
Formula Weight 152.58
Percent Purity 98%
Chemical Name or Material 2-Amino-6-chlorobenzonitrile
Show More Show Less
Hazard Category H317
Hazard Statement GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 229-501-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only