missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloro-3-nitropyridine, 98%

CAS: 27048-04-0 | C5H4ClN3O2 | 173.56 g/mol

$145.81 - $473.10

Chemical Identifiers

CAS 27048-04-0
Molecular Formula C5H4ClN3O2
Molecular Weight (g/mol) 173.56
MDL Number MFCD00044350
InChI Key WERABQRUGJIMKQ-UHFFFAOYSA-N
Synonym 2-amino-6-chloro-3-nitropyridine, 2-amino-3-nitro-6-chloropyridine, 6-chloro-3-nitro-2-pyridylamine, 6-chloro-3-nitro-2-pyridinamine, 6-amino-2-chloro-5-nitropyridine, 6-chloro-3-nitropyridin-2-ylamine, 6-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 6-chloro-3-nitro, 6-chloro-3-nitro-pyridin-2-amine, zlchem 113
PubChem CID 2724035
IUPAC Name 6-chloro-3-nitropyridin-2-amine
SMILES NC1=NC(Cl)=CC=C1[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6463414
View Documents
Thermo Scientific Chemicals
H6463414
25 g
Each for $145.81
Only null left
 
AAH6463422
View Documents
Thermo Scientific Chemicals
H6463422
100 g
Each for $473.10
Only null left
 
Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

27048-04-0
173.56
WERABQRUGJIMKQ-UHFFFAOYSA-N
2724035
NC1=NC(Cl)=CC=C1[N+]([O-])=O
C5H4ClN3O2
MFCD00044350
2-amino-6-chloro-3-nitropyridine, 2-amino-3-nitro-6-chloropyridine, 6-chloro-3-nitro-2-pyridylamine, 6-chloro-3-nitro-2-pyridinamine, 6-amino-2-chloro-5-nitropyridine, 6-chloro-3-nitropyridin-2-ylamine, 6-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 6-chloro-3-nitro, 6-chloro-3-nitro-pyridin-2-amine, zlchem 113
6-chloro-3-nitropyridin-2-amine

Specifications

27048-04-0
Yellow
MFCD00044350
UN2811
2-amino-6-chloro-3-nitropyridine, 2-amino-3-nitro-6-chloropyridine, 6-chloro-3-nitro-2-pyridylamine, 6-chloro-3-nitro-2-pyridinamine, 6-amino-2-chloro-5-nitropyridine, 6-chloro-3-nitropyridin-2-ylamine, 6-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 6-chloro-3-nitro, 6-chloro-3-nitro-pyridin-2-amine, zlchem 113
WERABQRUGJIMKQ-UHFFFAOYSA-N
6-chloro-3-nitropyridin-2-amine
2724035
98%
2-Amino-6-chloro-3-nitropyridine
192°C to 196°C
C5H4ClN3O2
25 g
474607
Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform.
NC1=NC(Cl)=CC=C1[N+]([O-])=O
173.56
173.56
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 248-188-5

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only