missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-bromopurine, 98%

Catalog No. AAB2162903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2162903 1 g
AAB2162906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2162903 Supplier Thermo Scientific Chemicals Supplier No. B2162903

CAS: 82499-03-4 | C5H4BrN5 | 214.026 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82499-03-4
Molecular Formula C5H4BrN5
Molecular Weight (g/mol) 214.026
MDL Number MFCD01321309
InChI Key HPGBGNVPUMCKPM-UHFFFAOYSA-N
Synonym 2-amino-6-bromopurine, 6-bromo-9h-purin-2-amine, 1h-purin-2-amine, 6-bromo, 6-bromoguanine, d2-amino-6-bromopurine, 6-bromopurine-2-ylamine, purine, 2-amino-6-bromo, 6-bromo-9h-purin-2-ylamine, 9h-purin-2-amine,6-bromo
PubChem CID 6101139
IUPAC Name 6-bromo-7H-purin-2-amine
SMILES C1=NC2=C(N1)C(=NC(=N2)N)Br

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 214.02
Percent Purity 98%
Chemical Name or Material 2-Amino-6-bromopurine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only