missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-bromobenzothiazole, 98%

Catalog No. p-7024228
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1855403 1 g
AAA1855406 5 g
AAA1855414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1855403 Supplier Thermo Scientific Chemicals Supplier No. A1855403
Only null left

CAS: 15864-32-1 | C7H5BrN2S | 229.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15864-32-1
Molecular Formula C7H5BrN2S
Molecular Weight (g/mol) 229.10
MDL Number MFCD00152229
InChI Key VZEBSJIOUMDNLY-UHFFFAOYSA-N
Synonym 2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304
PubChem CID 85149
IUPAC Name 6-bromo-1,3-benzothiazol-2-amine
SMILES NC1=NC2=CC=C(Br)C=C2S1

Specifications

Odor Odorless
Quantity 1 g
Formula Weight 229.1
Percent Purity 98%
Chemical Name or Material 2-Amino-6-bromobenzothiazole
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only