missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-trifluoromethyl-1,3,4-thiadiazole 98.0+%, TCI America™
$126.76 - $445.51
Chemical Identifiers
| CAS | 10444-89-0 |
|---|---|
| Molecular Formula | C3H2F3N3S |
| Molecular Weight (g/mol) | 169.125 |
| MDL Number | MFCD00003109 |
| InChI Key | LTEUXHSAYOSFGQ-UHFFFAOYSA-N |
| Synonym | 5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-amino-5-trifluoromethyl-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine, 5-trifluoromethyl, 5-trifluoromethyl-1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazole, 2-amino-5-trifluoromethyl, 5-trifluoromethyl-1,3,4-thiadiazole-2-ylamine, zlchem 1059, pubchem14503, maybridge1_003448 |
| PubChem CID | 82634 |
| IUPAC Name | 5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine |
| SMILES | C1(=NN=C(S1)N)C(F)(F)F |
Chemical Identifiers
| 10444-89-0 | |
| 169.125 | |
| LTEUXHSAYOSFGQ-UHFFFAOYSA-N | |
| 82634 | |
| C1(=NN=C(S1)N)C(F)(F)F |
| C3H2F3N3S | |
| MFCD00003109 | |
| 5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-amino-5-trifluoromethyl-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine, 5-trifluoromethyl, 5-trifluoromethyl-1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazole, 2-amino-5-trifluoromethyl, 5-trifluoromethyl-1,3,4-thiadiazole-2-ylamine, zlchem 1059, pubchem14503, maybridge1_003448 | |
| 5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine |
Specifications
| 10444-89-0 | |
| White-Yellow | |
| MFCD00003109 | |
| 5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-amino-5-trifluoromethyl-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine, 5-trifluoromethyl, 5-trifluoromethyl-1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazole, 2-amino-5-trifluoromethyl, 5-trifluoromethyl-1,3,4-thiadiazole-2-ylamine, zlchem 1059, pubchem14503, maybridge1_003448 | |
| C1(=NN=C(S1)N)C(F)(F)F | |
| 169.125 | |
| 169.13 | |
| Crystalline Powder |
| 223°C | |
| C3H2F3N3S | |
| 5 g | |
| LTEUXHSAYOSFGQ-UHFFFAOYSA-N | |
| 5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine | |
| 82634 | |
| ≥98.0% (HPLC) | |
| 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole |
Safety and Handling
TSCA : Yes